COM448618453 Combi-Blocks, Inc.

4-BOC-2,3-Hihydro-1,4-benzoxazine-6-boronic acid

  • MDL number MFCD20441846
  • Cas Number : 2096340-12-2
  • Molecular Weight : 279.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(tert-butoxycarbonyl)-3,4-dihydro-2H-1,4-benzoxazin-6-ylboronic acid
Inchi Code 1S/C13H18BNO5/c1-13(2,3)20-12(16)15-6-7-19-11-5-4-9(14(17)18)8-10(11)15/h4-5,8,17-18H,6-7H2,1-3H3
Inchi Key TWQCRBQJOUVYBQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-7444

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-7444
Availability details for
Quantity
  • Availability
  • Add to Cart