COM448618468 Combi-Blocks, Inc.

3-Borono-a,a-dimethyl-benzeneacetic acid

  • MDL number MFCD16652406
  • Cas Number : 885068-00-8
  • Molecular Weight : 208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-0896

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-0896
Availability details for
Quantity
  • Availability
  • Add to Cart