COM448618488 Combi-Blocks, Inc.

2-(Trifluoromethyl)-1h-benzimidazole-5-boronic acid, HCl salt

  • MDL number MFCD17167295
  • Cas Number : 2096332-98-6
  • Molecular Weight : 266.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1179

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1179
Availability details for
Quantity
  • Availability
  • Add to Cart