COM448618491 Combi-Blocks, Inc.

1-Boc-3-trifluoromethylpyrazole-5-boronic acid

  • MDL number MFCD17214177
  • Cas Number : 1684433-60-0
  • Molecular Weight : 280

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1192

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1192
Availability details for
Quantity
  • Availability
  • Add to Cart