COM448618493 Combi-Blocks, Inc.

1-[2-(4-Boronophenoxy)ethyl]-1,4-dihydropyridin-4-one

  • MDL number MFCD17402509
  • Cas Number : 1615247-97-6
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1250

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1250
Availability details for
Quantity
  • Availability
  • Add to Cart