COM448618495 Combi-Blocks, Inc.

2-Cyano-3-(trifluoromethyl)phenylboronic acid

  • MDL number MFCD17676534
  • Cas Number : 1198181-40-6
  • Molecular Weight : 214.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1326
Availability details for
Quantity
  • Availability
  • Add to Cart