COM448618508 Combi-Blocks, Inc.

(2,5-dimethoxypyridin-4-yl)boronic acid

  • MDL number MFCD18256800
  • Cas Number : 1630193-77-9
  • Molecular Weight : 183

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1724

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FB-1724
Availability details for
Quantity
  • Availability
  • Add to Cart