COM448618522 Combi-Blocks, Inc.

5-(Benzyloxy)-2-formylphenylboronic acid

  • MDL number MFCD18383732
  • Cas Number : 1226773-36-9
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2484
Availability details for
Quantity
  • Availability
  • Add to Cart