COM448618525 Combi-Blocks, Inc.

2-Methoxyquinolin-6-ylboronic acid

  • MDL number MFCD18384109
  • Cas Number : 1191061-58-1
  • Molecular Weight : 203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2565

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2565
Availability details for
Quantity
  • Availability
  • Add to Cart