COM448618526 Combi-Blocks, Inc.

2,3-Dichloro-4-methoxyphenylboronic acid

  • MDL number MFCD18384131
  • Cas Number : 1190219-72-7
  • Molecular Weight : 220.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2585

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2585
Availability details for
Quantity
  • Availability
  • Add to Cart