COM448618529 Combi-Blocks, Inc.

2-Chloro-6-(trifluoromethoxy)phenylboronic acid

  • MDL number MFCD18393880
  • Cas Number : 2036084-49-6
  • Molecular Weight : 240.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2657

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2657
Availability details for
Quantity
  • Availability
  • Add to Cart