COM448618539 Combi-Blocks, Inc.

1-Butylpyrazole-4-boronic acid

  • MDL number MFCD20441847
  • Cas Number : 2096331-96-1
  • Molecular Weight : 168.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-5394

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-5394
Availability details for
Quantity
  • Availability
  • Add to Cart