COM448618540 Combi-Blocks, Inc.

3,4-Difluoro-5-hydroxyphenylboronic acid

  • MDL number MFCD22414421
  • Cas Number : 1379466-84-8
  • Molecular Weight : 173.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-5473

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-5473
Availability details for
Quantity
  • Availability
  • Add to Cart