COM448618544 Combi-Blocks, Inc.

2-Isopropoxy-5-methylpyridine-3-boronic acid

  • MDL number MFCD19237189
  • Cas Number : 2096337-70-9
  • Molecular Weight : 195.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-6173

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-6173
Availability details for
Quantity
  • Availability
  • Add to Cart