COM448618545 Combi-Blocks, Inc.

8-Fluoro-2,3-dihydro-1,4-benzoxazine-6-boronic acid

  • MDL number MFCD20441849
  • Cas Number : 1701449-28-6
  • Molecular Weight : 197.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-6269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-6269
Availability details for
Quantity
  • Availability
  • Add to Cart