COM448618546 Combi-Blocks, Inc.

2-(Azepan-1-yl)pyrimidine-5-boronic acid

  • MDL number MFCD19684111
  • Cas Number : 2096333-81-0
  • Molecular Weight : 221.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-7340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-7340
Availability details for
Quantity
  • Availability
  • Add to Cart