COM448618550 Combi-Blocks, Inc.

1-BOC-1,2,3,4-Tetrahydroquinoline-7-boronic acid

  • MDL number MFCD20441850
  • Cas Number : 1926137-46-3
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-8918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-8918
Availability details for
Quantity
  • Availability
  • Add to Cart