COM448618553 Combi-Blocks, Inc.

2,5-Difluoropyridine-4-boronic acid, pinacol estser

  • MDL number MFCD18712644
  • Cas Number : 1622217-35-9
  • Molecular Weight : 241.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-0742

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-0742
Availability details for
Quantity
  • Availability
  • Add to Cart