COM448618570 Combi-Blocks, Inc.

4-Fluoro-2-methoxy-5-methylphenylboronic acid, pinacol ester

  • MDL number MFCD19981541
  • Cas Number : 2096334-36-8
  • Molecular Weight : 266.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-2523

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-2523
Availability details for
Quantity
  • Availability
  • Add to Cart