COM448618573 Combi-Blocks, Inc.

4-Methylpyrazole-5-boronic acid pinacol ester

  • MDL number MFCD18250651
  • Cas Number : 1440520-87-5
  • Molecular Weight : 208.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5027

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5027
Availability details for
Quantity
  • Availability
  • Add to Cart