COM448618589 Combi-Blocks, Inc.

1-Butyl-1H-pyrazole-4-boronic acid, pinacol ester

  • MDL number MFCD17014638
  • Cas Number : 1488423-70-6
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5394

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5394
Availability details for
Quantity
  • Availability
  • Add to Cart