COM448618593 Combi-Blocks, Inc.

3-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-indole

  • MDL number MFCD16995768
  • Cas Number : 1072812-35-1
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5485

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5485
Availability details for
Quantity
  • Availability
  • Add to Cart