COM448618603 Combi-Blocks, Inc.

2-Hydroxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

  • MDL number MFCD16994242
  • Cas Number : 741698-75-9
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5694
Availability details for
Quantity
  • Availability
  • Add to Cart