COM448618606 Combi-Blocks, Inc.

3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile

  • MDL number MFCD16995199
  • Cas Number : 1220219-63-5
  • Molecular Weight : 264.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5725

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5725
Availability details for
Quantity
  • Availability
  • Add to Cart