COM448618608 Combi-Blocks, Inc.

3-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazol-1-yl)propanamide

  • MDL number MFCD16660238
  • Cas Number : 1093307-34-6
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5733
Availability details for
Quantity
  • Availability
  • Add to Cart