COM448618619 Combi-Blocks, Inc.

1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-indazol-3-amine

  • MDL number MFCD18383794
  • Cas Number : 1220220-18-7
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5931

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FF-5931
Availability details for
Quantity
  • Availability
  • Add to Cart