COM448618620 Combi-Blocks, Inc.

4-Methyl-2,3-dihydrobenzo-1,4-oxazine-6-boronic acid, pinacol ester

  • MDL number MFCD18383442
  • Cas Number : 1256256-24-2
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6044

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6044
Availability details for
Quantity
  • Availability
  • Add to Cart