COM448618626 Combi-Blocks, Inc.

Methyl 2-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number MFCD18384010
  • Cas Number : 1198615-60-9
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6153
Availability details for
Quantity
  • Availability
  • Add to Cart