COM448618632 Combi-Blocks, Inc.

Methyl 3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazol-1-yl)propanoate

  • MDL number MFCD16660987
  • Cas Number : 1093307-33-5
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6287

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6287
Availability details for
Quantity
  • Availability
  • Add to Cart