COM448618635 Combi-Blocks, Inc.

1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-indazole-5-carbonitrile

  • MDL number MFCD16995856
  • Cas Number : 2377609-00-0
  • Molecular Weight : 283.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6377
Availability details for
Quantity
  • Availability
  • Add to Cart