COM448618638 Combi-Blocks, Inc.

4-Methoxyquinoline-7-boronic acid, pinacol ester

  • MDL number MFCD18383860
  • Cas Number : 2377607-98-0
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6432

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6432
Availability details for
Quantity
  • Availability
  • Add to Cart