COM448618639 Combi-Blocks, Inc.

2-(4-Methylimidazol-1-yl)pyrimidine-5-boronic acid, pinacol ester

  • MDL number MFCD16618881
  • Cas Number : 1402240-42-9
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6461

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6461
Availability details for
Quantity
  • Availability
  • Add to Cart