COM448618640 Combi-Blocks, Inc.

5-Methoxy-1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-indazole

  • MDL number MFCD16995854
  • Cas Number : 2377609-98-6
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6599

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6599
Availability details for
Quantity
  • Availability
  • Add to Cart