COM448618660 Combi-Blocks, Inc.

2-Isopropoxy-3-(trifluoromethyl)pyridine-5-boronic acid pinacol ester

  • MDL number MFCD16996732
  • Cas Number : 1334607-81-6
  • Molecular Weight : 331.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8298
Availability details for
Quantity
  • Availability
  • Add to Cart