COM448618670 Combi-Blocks, Inc.

1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)-indazol-1-yl)ethanone

  • MDL number MFCD16995895
  • Molecular Weight : 354.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8812

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8812
Availability details for
Quantity
  • Availability
  • Add to Cart