COM448618688 Combi-Blocks, Inc.

1-(Phenylsulfonyl)indole-6-boronnic acid pinacol ester

  • MDL number MFCD12828207
  • Cas Number : 1333344-24-3
  • Molecular Weight : 383.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1082

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1082
Availability details for
Quantity
  • Availability
  • Add to Cart