COM448618691 Combi-Blocks, Inc.

5-Bromobenzofuran-7-boronic acid, pinacol ester

  • MDL number MFCD18783161
  • Cas Number : 2096333-86-5
  • Molecular Weight : 325.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1132
Availability details for
Quantity
  • Availability
  • Add to Cart