COM448618692 Combi-Blocks, Inc.

4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)butan-2-one

  • MDL number MFCD22208512
  • Cas Number : 100818-32-4
  • Molecular Weight : 198.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1280

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1280
Availability details for
Quantity
  • Availability
  • Add to Cart