COM448618696 Combi-Blocks, Inc.

3-Methyl-5-trifluoromethylphenylboronic acid, pinacol ester

  • MDL number MFCD17926495
  • Cas Number : 1256358-84-5
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1308

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1308
Availability details for
Quantity
  • Availability
  • Add to Cart