COM448618700 Combi-Blocks, Inc.

2-(2,2,2-trifluoroethoxy)pyrimidine-5-boronic acid, pinacol ester

  • MDL number MFCD18311855
  • Cas Number : 1351234-12-2
  • Molecular Weight : 304.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1448

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1448
Availability details for
Quantity
  • Availability
  • Add to Cart