COM448618703 Combi-Blocks, Inc.

3-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl chloride

  • MDL number MFCD20489143
  • Cas Number : 1181329-14-5
  • Molecular Weight : 266.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1497

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1497
Availability details for
Quantity
  • Availability
  • Add to Cart