COM448618708 Combi-Blocks, Inc.

3-Methoxy-2-(methoxycarbonyl)phenylboronic acid, pinacol ester

  • MDL number MFCD19684112
  • Cas Number : 1146214-77-8
  • Molecular Weight : 292.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Inchi Code 1S/C15H21BO5/c1-14(2)15(3,4)21-16(20-14)10-8-7-9-11(18-5)12(10)13(17)19-6/h7-9H,1-6H3
Inchi Key RAGDNDSTZKPURP-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1548
Availability details for
Quantity
  • Availability
  • Add to Cart