COM448618723 Combi-Blocks, Inc.

2-(2,5-Dimethylthiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD12405471
  • Cas Number : 942070-20-4
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1646

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1646
Availability details for
Quantity
  • Availability
  • Add to Cart