COM448618727 Combi-Blocks, Inc.

5-(2-Hydroxypropan-2-yl)pyridine-3-boronic acid pinacol ester

  • MDL number MFCD15143417
  • Cas Number : 1257431-63-2
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1750

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1750
Availability details for
Quantity
  • Availability
  • Add to Cart