COM448618738 Combi-Blocks, Inc.

3,5-Dimethyl-4-methoxycarbonylphenylboronic acid, pinacol ester

  • MDL number MFCD18311856
  • Cas Number : 2096333-27-4
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1919
Availability details for
Quantity
  • Availability
  • Add to Cart