COM448618740 Combi-Blocks, Inc.

3-(Cyclopropylsulfonyl)phenylboronic acid, pinacol ester

  • MDL number MFCD19237190
  • Cas Number : 1020206-37-4
  • Molecular Weight : 308.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2055

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2055
Availability details for
Quantity
  • Availability
  • Add to Cart