COM448618746 Combi-Blocks, Inc.

Methyl 2-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]acetate

  • MDL number MFCD24448779
  • Cas Number : 2377608-13-2
  • Molecular Weight : 292.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2204

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2204
Availability details for
Quantity
  • Availability
  • Add to Cart