COM448618747 Combi-Blocks, Inc.

4,4,5,5-tetramethyl-2-[(E)-2-[2-(trifluoromethyl)phenyl]ethenyl]-1,3,2-dioxaborolane

  • MDL number MFCD19703868
  • Cas Number : 1294009-25-8
  • Molecular Weight : 298.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2229

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2229
Availability details for
Quantity
  • Availability
  • Add to Cart