COM448618752 Combi-Blocks, Inc.

3,5-Difluoropyridine-4-boronic acid pinacol ester

  • MDL number MFCD12198149
  • Cas Number : 1310404-59-1
  • Molecular Weight : 241.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2302
Availability details for
Quantity
  • Availability
  • Add to Cart