COM448618768 Combi-Blocks, Inc.

4-Carboxy-3-methylphenylboronic acid, pinacol ester

  • MDL number MFCD09037836
  • Cas Number : 890839-22-2
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-2344
Availability details for
Quantity
  • Availability
  • Add to Cart